missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human ELAV-like protein 1

A DNA sequence encoding the Homo sapiens (Human) ELAV-like protein 2, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP5980-ye-50ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15919119

  • 310.00€ / 50 microgramos


Fecha estimada de envío
16-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_001164666.1
ELAV-like protein 1
50 μg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
1993
Greater than 90% as determined by SDS-PAGE.
Research Use Only
ELAVL2
Recombinant Protein
Yeast
QLSNGPTCNN TANGPTTINN NCSSPVDSGN TEDSKTNLIV NYLPQNMTQE ELKSLFGSIG EIESCKLVRD KITGQSLGYG FVNYIDPKDA EKAINTLNGL RLQTKTIKVS YARPSSASIR DANLYVSGLP KTMTQKELEQ LFSQYGRIIT SRILVDQVTG ISRGVGFIRF DKRIEAEEAI KGLNGQKPPG ATEPITVKFA NNPSQKTNQA ILSQLYQSPN RRYPGPLAQQ AQRFRLDNLL NMAYGVKRFS PMTIDGMTSL AGINIPGHPG TGWCIFVYNL APDADESILW QMFGPFGAVT NVKVIRDFNT NKCKGFGFVT MTNYDEAAMA IASLNGYRLG DRVLQVSFKT NKT
Videos