missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human EIF4EBP2 Protein

A DNA sequence encoding the Homo sapiens (Human) EIF4EBP2, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP5973-ye-500ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15909059

  • 1453.00€ / 500 microgramos


Fecha estimada de envío
14-08-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_004087.1
EIF4EBP2 Protein
500 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
1979
Greater than 90% as determined by SDS-PAGE.
Research Use Only
EIF4EBP2
Recombinant Protein
Yeast
MSSSAGSGHQ PSQSRAIPTR TVAISDAAQL PHDYCTTPGG TLFSTTPGGT RIIYDRKFLL DRRNSPMAQT PPCHLPNIPG VTSPGTLIED SKVEVNNLNN LNNHDRKHAV GDDAQFEMDI
Videos
Documentos