missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human EGF / Epidermal Growth Factor Protein
Recombinant Protein
Marca: enQuireBio™ QP5334-5mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001171601.1 | |
EGF / Epidermal Growth Factor Protein | |
Research Use Only | |
EGF | |
Recombinant Protein | |
E. coli | |
NSDSECPLSH . DGYCLHDGV C. MYIEALDK YA. CNCVVGY IGE. RCQYRD LKWW. ELR | |
Greater than 95% as determined by SDS-PAGE. |
1950 | |
5 mg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Shows bioactivity at a concentration of 100 pg/mL in the 3T3 cell proliferation assay. The bioactivity acceptance criteria is set at 1 x 10^7 units/mg. | |
Human | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |