missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human EFNB3 Protein
A cDNA sequence encoding the EFNB3 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11756-HIS-20ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
EFNB3 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSHMLSLEP VYWNSANKRF QAEGGYVLYP QIGDRLDLLC PRARPPGPHS SPNYEFYKLY LVGGAQGRRC EAPPAPNLLL TCDRPDLDLR FTIKFQEYSP NLWGHEFRSH HDYYIIATSD GTREGLESLQ GGVCLTRGMK VLLRVGQSPR GGAVPRKPVS EMPMERDRGA AHSLEPGKEN LPGDPTSNAT SRGAEGPLPP PSMP | |
Greater than 90% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
EFNB3 protein solution (1 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 20% glycerol, 0.1M NaCl and 2M urea. |