missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human EEF1E1 / AIMP3 / p18 Protein
A DNA sequence encoding the Homo sapiens (Human) EEF1E1 / AIMP3 / p18, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5961-ec-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001129122.1 | |
EEF1E1 / AIMP3 / p18 Protein | |
Research Use Only | |
EEF1E1 | |
Recombinant Protein | |
E. coli | |
AAAAELSLLE KSLGLSKGNK YSAQGERQIP VLQTNNGPSL TGLTTIAAHL VKQANKEYLL GSTAEEKAIV QQWLEYRVTQ VDGHSSKNDI HTLLKDLNSY LEDKVYLTGY NFTLADILLY YGLHRFIVDL TVQEKEKYLN VSRWFCHIQH YPGIRQHLSS VVFIKNRLYT NSH | |
Greater than 90% as determined by SDS-PAGE. |
9521 | |
500 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |