missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human EBNA1BP2 Protein
A cDNA sequence encoding the EBNA1BP2 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11725-20ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
EBNA1BP2 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMDTPPLS DSESESDESL VTDRELQDAF SRGLLKPGLN VVLEGPKKAV NDVNGLKQCL AEFKRDLEWV ERLDVTLGPV PEIGGSEAPA PQNKDQKAVD PEDDFQREMS FYRQAQAAVL AVLPRLHQLK VPTKRPTDYF AEMAKSDLQM QKIRQKLQTK QAAMERSEKA KQLRALRKYG KKVQTEVLQK RQQEKAHMMN AIKKYQKGFS DKLDFLEGDQ KPLAQRKKAG AKGQQMRKGP SAKRRYKNQK FGFGGKKKGS KWNTRESYDD VSSFRAKTAH GRGLKRPGKK GSNKRPGKRT REKMKNRTH | |
Greater than 80% as determined by SDS-PAGE. |
20 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The EBNA1BP2 solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol, 0.4M Urea. |