missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human E3 ubiquitin-protein ligase RFWD2 Protein
A DNA sequence encoding the Homo sapiens(Human) E3 ubiquitin-protein ligase RFWD2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10010-iv-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001001740.1 | |
E3 ubiquitin-protein ligase RFWD2 Protein | |
Research Use Only | |
RFWD2 | |
Recombinant Protein | |
E. coli | |
MSGSRQAGSG SAGTSPGSSA ASSVTSASSS LSSSPSPPSV AVSAAALVSG GVAQAAGSGG LGGPVRPVLV APAVSGSGGG AVSTGLSRHS CAARPSAGVG GSSSSLGSGS RKRPLLAPLC NGLINSYEDK SNDFVCPICF DMIEEAYMTK CGHSFCYKCI HQSLEDNNRC PKCNYVVDNI DHLYPNFLVN ELILKQKQRF EEKRFKLDHS VSSTNGHRWQ IFQDWLGTDQ DNLDLANVNL MLELLVQKKK QLEAESHAAQ LQILMEFLKV ARRNKREQLE QIQKELSVLE EDIKRVEEMS GLYSPVSEDS TVPQFEAPSP SHSSIIDSTE YSQPPGFSGS SQTKKQPWYN STLASRRKRL TAHFEDLEQC YFSTRMSRIS DDSRTASQLD EFQECLSKFT RYNSVRPLAT LSYASDLYNG SSIVSSIEFD RDCDYFAIAG VTKKIKVYEY DTVIQDAVDI HYPENEMTCN SKISCISWSS YHKNLLASSD YEGTVILWDG FTGQRSKVYQ EHEKRCWSVD FNLMDPKLLA SGSDDAKVKL WSTNLDNSVA SIEAKANVCC VKFSPSSRYH LAFGCADHCV HYYDLRNTKQ PIMVFKGHRK AVSYAKFVSG EEIVSASTDS QLKLWNVGKP YCLRSFKGHI NEKNFVGLAS NGDYIACGSE NNSLYLYYKG LSKTLLTFKF DTVKSVLDKD RKEDDTNEFV SAVCWRALPD GESNVLIAAN SQGTIKVLEL V | |
Greater than 85% as determined by SDS-PAGE. |
64326 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |