missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human DUSP21 Protein

A cDNA sequence encoding the DUSP21 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP11714-1mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15908128

  • 5941.00€ / 1 miligramo


Fecha estimada de envío
22-08-2019
Añadir a la cesta

Especificaciones

Especificaciones

DUSP21 Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The DUSP21 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 200mM NaCl, 2mM DTT and 20% glycerol.
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMTASAS SFSSSQGVQQ PSIYSFSQIT RSLFLSNGVA ANDKLLLSSN RITAIVNASV EVVNVFFEGI QYIKVPVTDA RDSRLYDFFD PIADLIHTID MRQGRTLLHC MAGVSRSASL CLAYLMKYHS MSLLDAHTWT KSRRPIIRPN NGFWEQLINY EFKLFNNNTV RMINSPVGNI PDIYEKDLRM MISM
Videos
Documentos