missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DUSP14 / MKP-6 Protein
A DNA sequence encoding the Homo sapiens (Human) DUSP14 / MKP-6, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5957-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
DUSP14 / MKP-6 Protein | |
Research Use Only | |
DUSP14 | |
Recombinant Protein | |
E. coli | |
MSSRGHSTLP RTLMAPRMIS EGDIGGIAQI TSSLFLGRGS VASNRHLLQA RGITCIVNAT IEIPNFNWPQ FEYVKVPLAD MPHAPIGLYF DTVADKIHSV SRKHGATLVH CAAGVSRSAT LCIAYLMKFH NVCLLEAYNW VKARRPVIRP NVGFWRQLID YERQLFGKST VKMVQTPYGI VPDVYEKESR HLMPYWGI | |
Greater than 90% as determined by SDS-PAGE. |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |