missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DTD1 Protein
A cDNA sequence encoding the DTD1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11708-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
92675 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSMKAVVQR VTRASVTVGG EQISAIGRGI CVLLGISLED TQKELEHMVR KILNLRVFED ESGKHWSKSVMDKQYEILCV SQFTLQCVLK GNKPDFHLAM PTEQAEGFYN SFLEQLRKTY RPELIKDGKF GAYMQVHIQN DGPVTIELES PAPGTATSDP KQLSKLEKQQ QRKEKTRAKG PSESSKERNT PRKEDRSASS GAEGDVSSER EP | |
Greater than 85% as determined by SDS-PAGE. |
DTD1 Protein | |
Research Use Only | |
DTD1 | |
Human | |
His | |
DTD1 protein solution (0.5 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.1M NaCl, 10% glycerol and 1mM DTT. |