missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human DNTTIP1 Protein

A DNA sequence encoding the Homo sapiens (Human) DNTTIP1, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP5942-ec-10ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15918839

  • 183.00€ / 10 microgramos


Fecha estimada de envío
14-08-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_443183.1
DNTTIP1 Protein
10 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
116092
Greater than 90% as determined by SDS-PAGE.
Research Use Only
DNTTIP1
Recombinant Protein
E. coli
MGATGDAEQP RGPSGAERGG LELGDAGAAG QLVLTNPWNI MIKHRQVQRR GRRSQMTTSF TDPAISMDLL RAVLQPSINE EIQTVFNKYM KFFQKAALNV RDNVGEEVDA EQLIQEACRS CLEQAKLLFS DGEKVIPRLT HELPGIKRGR QAEEECAHRG SPLPKKRKGR PPGHILSSDR AAAGMVWKPK SCEPIRREGP KWDPARLNES TTFVLGSRAN KALGMGGTRG RIYIKHPHLF KYAADPQDKH WLAEQHHMRA TGGKMAYLLI EEDIRDLAAS DDYRGCLDLK LEELKSFVLP SWMVEKMRKY METLRTENEH RAVEAPPQT
Videos
Documentos