missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human DNAJC24 Protein

A cDNA sequence encoding the DNAJC24 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP11680-2ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15908028

  • 103.00€ / 2 microgramos


Fecha estimada de envío
22-08-2019
Añadir a la cesta

Especificaciones

Especificaciones

DNAJC24 Protein
2 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The DNAJC24 solution (1 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 10% glycerol and 1mM DTT.
Greater than 90% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSMMAVEQM PKKDWYSILG ADPSANISDL KQKYQKLILM YHPDKQSTDV PAGTVEECVQ KFIEIDQAWK ILGNEETKRE YDLQRCEDDL RNVGPVDAQV YLEEMSWNEG DHSFYLSCRC GGKYSVSKDE AEEVSLISCD TCSLIIELLH YN
Videos
Documentos