missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DNAJC15 Protein
A cDNA sequence encoding the DNAJC15 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11679-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
29103 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MGSFRIWKPL EQVITETAKK ISTPSFSSYY KGGFEQKMSR REAGLILGVS PSAGKAKIRT AHRRVMILNH PDKGGSPYVA AKINEAKDLL ETTTKH | |
Greater than 90% as determined by SDS-PAGE. |
DNAJC15 Protein | |
Research Use Only | |
DNAJC15 | |
Human | |
His | |
The DNAJC15 solution (0.25 mg/ml) contains 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 20% glycerol and 1mM DTT. |