missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DnaJ homolog subfamily B member 2 Protein
A DNA sequence encoding the Homo sapiens (Human) DnaJ homolog subfamily B member 2, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5938-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001034639.1 | |
DnaJ homolog subfamily B member 2 Protein | |
Research Use Only | |
DNAJB2 | |
Recombinant Protein | |
E. coli | |
ASYYEILDVP RSASADDIKK AYRRKALQWH PDKNPDNKEF AEKKFKEVAE AYEVLSDKHK REIYDRYGRE GLTGTGTGPS RAEAGSGGPG FTFTFRSPEE VFREFFGSGD PFAELFDDLG PFSELQNRGS RHSGPFFTFS SSFPGHSDFS SSSFSFSPGA GAFRSVSTST TFVQGRRITT RRIMENGQER VEVEEDGQLK SVTINGVPDD LALGLELSRR EQQPSVTSRS GGTQVQQTPA SCPLDSDLSE DEDLQLAMAY SLSEMEAAGK KPAGGREAQH RRQGRPKAQH QDPGLGGTQE GARGEATKRS PSPEEKASRC LIL | |
Greater than 90% as determined by SDS-PAGE. |
3300 | |
10 μg | |
Not determined. | |
Full Length of Isoform 3 | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |