missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human DNA-directed RNA polymerase III subunit RPC1 Protein

A DNA sequence encoding the Homo sapiens (Human) DNA-directed RNA polymerase III subunit RPC1, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP6511-ec-10ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16053820

  • 183.00€ / 10 microgramos


Fecha estimada de envío
13-11-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_008986.2
DNA-directed RNA polymerase III subunit RPC1 Protein
10 μg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
11128
Greater than 90% as determined by SDS-PAGE.
Research Use Only
POLR3A
Recombinant Protein
E. coli
FPEKVNKANI NFLRKLVQNG PEVHPGANFI QQRHTQMKRF LKYGNREKMA QELKYGDIVE RHLIDGDVVL FNRQPSLHKL SIMAHLARVK PHRTFRFNEC VCTPYNADFD GDEMNLHLPQ TEEAKAEALV LMGTKANLVT PRNGEPLIAA IQDFLTGAYL LTLKDTFFDR AKACQIIASI LVGKDEKIKV RLPPPTILKP VTLWTGKQIF SVILRPSDDN PVRANLRTKG KQYCGKGEDL C
Videos
Documentos