missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human DNA-directed RNA polymerase I subunit RPA12 Protein
A DNA sequence encoding the Homo sapiens (Human) DNA-directed RNA polymerase I subunit RPA12, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8203-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001265714.1 | |
DNA-directed RNA polymerase I subunit RPA12 Protein | |
Research Use Only | |
ZNRD1 | |
Recombinant Protein | |
E. coli | |
MSVMDLANTC SSFQSDLDFC SDCGSVLPLP GAQDTVTCIR CGFNINVRDF EGKVVKTSVV FHQLGTAMPM SVEEGPECQG PVVDRRCPRC GHEGMAYHTR QMRSADEGQT VFYTCTNCKF QEKEDS | |
Greater than 90% as determined by SDS-PAGE. |
30834 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |