missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human DLAT, mitochondrial Protein

A DNA sequence encoding the Homo sapiens (Human) DLAT, mitochondrial, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP5936-ec-1mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15918789

  • 1857.00€ / 1 miligramo


Fecha estimada de envío
16-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_001922.2
DLAT, mitochondrial Protein
1 mg
Not determined.
Partial (see sequence information for more details).
Human
His
Tris-based buffer, 50% glycerol
1737
Greater than 90% as determined by SDS-PAGE.
Research Use Only
DLAT
Recombinant Protein
E. coli
KVPLPSLSPT MQAGTIARWE KKEGDKINEG DLIAEVETDK ATVGFESLEE CYMAKILVAE GTRDVPIGAI ICITVGKPED IEAFKNYTLD SSAAPTPQAA PAPTPAATAS PPTPSAQAPG SSYPPHMQVL LPALSPTMTM GTVQRWEKKV GEKLSEGDLL AEIETDKATI GFEVQEEGYL AKILVPEGTR DVPLGTPLCI IVEKEADISA FADYRPTEVT DLKPQVPPPT PPPVAAVPPT PQPLAPTPSA PCPATPAGPK GRVFVSPLAK KLAVEKGIDL TQVKGTGPDG RITKKDIDSF VPSKVAPAPA AVVPPTGPGM APVPTGVFTD IPISNIRRVI AQRLMQSKQT IPHYYLSIDV NMGEVLLVRK ELNKILEGRS KISVNDFIIK ASALACLKVP EANSSWMDTV IRQNHVVDVS VAVSTPAGLI TPIVFNAHIK GVETIANDVV SLATKAREGK LQPHEFQGGT FTISNLGMFG IKNFSAIINP PQACILAIGA SEDKLVPADN EKGFDVASMM SVTLSCDHRV VDGAVGAQWL AEFRKYLE
Videos