missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Diacylglycerol kinase gamma Protein

A DNA sequence encoding the Homo sapiens (Human) Diacylglycerol kinase gamma, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP5931-ec-200ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15928759

  • 675.00€ / 200 microgramos


Fecha estimada de envío
20-03-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_001074213.1
Diacylglycerol kinase gamma Protein
200 μg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
1608
Greater than 90% as determined by SDS-PAGE.
Research Use Only
DGKG
Recombinant Protein
E. coli
MGEERWVSLT PEEFDQLQKY SEYSSKKIKD ALTEFNEGGS LKQYDPHEPI SYDVFKLFMR AYLEVDLPQP LSTHLFLAFS QKPRHETSDH PTEGASNSEA NSADTNIQNA DNATKADEAC APDTESNMAE KQAPAEDQVA ATPLEPPVPR SSSSESPVVY LKDVVCYLSL LETGRPQDKL EFMFRLYDSD ENGLLDQAEM DCIVNQMLHI AQYLEWDPTE LRPILKEMLQ GMDYDRDGFV SLQEWVHGGM TTIPL
Videos
Documentos