missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Death domain-associated protein 1
A DNA sequence encoding the Homo sapiens (Human) Death domain-associated protein 6, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8355-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001135441.1 | |
Death domain-associated protein 1 | |
Research Use Only | |
DAXX | |
Recombinant Protein | |
E. coli | |
MATANSIIVL DDDDEDEAAA QPGPSHPLPN AASPGAEAPS SSEPHGARGS SSSGGKKCYK LENEKLFEEF LELCKMQTAD HPEVVPFLYN RQQRAHSLFL ASAEFCNILS RVLSRARSRP AKLYVYINEL CTVLKAHSAK KKLNLAPAAT TSNEPSGNNP PTHLSLDPTN AENTASQSPR TRGSRRQIQR LEQLLALYVA EIRRLQEKEL DLSELDDPDS AYLQEARLKR KLI | |
Greater than 90% as determined by SDS-PAGE. |
1616 | |
10 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |