missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human DCK Protein

A cDNA sequence encoding the DCK was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP11599-5ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15907768

  • 103.00€ / 5 microgramos


Fecha de envío estimada
del proveedor 21-05-2019
Añadir a la cesta

Especificación

Especificación

DCK Protein
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The DCK solution (0.5 mg/ml) contains 20mM Tris-HCl Buffer (pH 7.5), 1mM DTT, 0.1mM PMSF, 2mM EDTA and 10% Glycerol.
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MRGSHHHHHH GMASMTGGQQ MGRDLYDDDD KDRWGSMATP PKRSCPSFSA SSEGTRIKKI SIEGNIAAGK STFVNILKQL CEDWEVVPEP VARWCNVQST QDEFEELTMS QKNGGNVLQM MYEKPERWSF TFQTYACLSR IRAQLASLNG KLKDAEKPVL FFERSVYSDR YIFASNLYES ECMNETEWTI YQDWHDWMNN QFGQSLELDG IIYLQATPET CLHRIYLRGR NEEQGIPLEY LEKLHYKHES WLLHRTLKTN FDYLQEVPIL TLDVNEDFKD KYESLVEKVK EFLSTL
Videos