missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human d2hgdh, mitochondrial Protein
A DNA sequence encoding the Homo sapiens (Human) d2hgdh, mitochondrial, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP7919-ec-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001274178.1 | |
d2hgdh, mitochondrial Protein | |
Research Use Only | |
d2hgdh | |
Recombinant Protein | |
E. coli | |
LRGAPGAAGS WGRPVGPLAR RGCCSAPGTP EVPLTRERYP VRRLPFSTVS KQDLAAFERI VPGGVVTDPE ALQAPNVDWL RTLRGCSKVL LRPRTSEEVS HILRHCHERN LAVNPQGGNT GMVGGSVPVF DEIILSTARM NRVLSFHSVS GILVCQAGCV LEELSRYVEE RDFIMPLDLG AKGSCHIGGN VATNAGGLRF LRYGSLHGTV LGLEVVLADG TVLDCLTSLR KDNTGYDLKQ LFIGSEGTLG IITTVSILCP PKPRAVNVAF LGCPGFAEVL QTFSTCKGML GEILSAFEFM DAVCMQLVGR HLHLASPVQE SPFYVLIETS GSNAGHDAEK LGHFLEHALG SGLVTDGTMA TDQRKVKMLW ALRERITEAL SRDGYVYKYD LSLPVERLYD IVTDLRARLG PHAKHVVGYG HLGDGNLHLN VTAEAFSPSL LAALEPHVYE WTAGQQGSVS AEHGVGFRKR DVLGYSKPPG ALQLMQQLKA LLDPKGILNP YKTLPSQA | |
Greater than 90% as determined by SDS-PAGE. |
728294 | |
10 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |