missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Cytochrome P450 11B2, mitochondrial Protein

A DNA sequence encoding the Homo sapiens (Human) Cytochrome P450 11B2, mitochondrial, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP5903-ec-10ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15928559

  • 245.00€ / 10 microgramos


Fecha estimada de envío
01-04-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_000489.3
Cytochrome P450 11B2, mitochondrial Protein
10 μg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
1585
Greater than 90% as determined by SDS-PAGE.
Research Use Only
CYP11B2
Recombinant Protein
E. coli
GTRAARAPRT VLPFEAMPQH PGNRWLRLLQ IWREQGYEHL HLEMHQTFQE LGPIFRYNLG GPRMVCVMLP EDVEKLQQVD SLHPCRMILE PWVAYRQHRG HKCGVFLLNG PEWRFNRLRL NPDVLSPKAV QRFLPMVDAV ARDFSQALKK KVLQNARGSL TLDVQPSIFH YTIEASNLAL FGERLGLVGH SPSSASLNFL HALEVMFKST VQLMFMPRSL SRWISPKVWK EHFEAWDCIF QYGDNCIQKI YQELAFNRPQ HYTGIVAELL LKAELSLEAI KANSMELTAG SVDTTAFPLL MTLFELARNP DVQQILRQES LAAAASISEH PQKATTELPL LRAALKETLR LYPVGLFLER VVSSDLVLQN YHIPAGTLVQ VFLYSLGRNA ALFPRPERYN PQRWLDIRGS GRNFHHVPFG FGMRQCLGRR LAEAEMLLLL HHVLKHFLVE TLTQEDIKMV YSFILRPGTS PLLTFRAIN
Videos
Documentos