missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Cytochrome b-245 heavy chain Protein

A DNA sequence encoding the Homo sapiens (Human) Cytochrome b-245 heavy chain, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP5899-ec-50ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15928539

  • 253.00€ / 50 microgramos


Fecha estimada de envío
21-10-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_000388.2
Cytochrome b-245 heavy chain Protein
50 μg
Not determined.
Cytoplasmic Domain
Human
His-SUMO
Tris-based buffer, 50% glycerol
1536
Greater than 90% as determined by SDS-PAGE.
Research Use Only
CYBB
Recombinant Protein
E. coli
ERLVRFWRSQ QKVVITKVVT HPFKTIELQM KKKGFKMEVG QYIFVKCPKV SKLEWHPFTL TSAPEEDFFS IHIRIVGDWT EGLFNACGCD KQEFQDAWKL PKIAVDGPFG TASEDVFSYE VVMLVGAGIG VTPFASILKS VWYKYCNNAT NLKLKKIYFY WLCRDTHAFE WFADLLQLLE SQMQERNNAG FLSYNIYLTG WDESQANHFA VHHDEEKDVI TGLKQKTLYG RPNWDNEFKT IASQHPNTRI GVFLCGPEAL AETLSKQSIS NSESGPRGVH FIFNKENF
Videos
Documentos