missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Cysteine and glycine-rich protein 1

A DNA sequence encoding the Homo sapiens (Human) Cysteine and glycine-rich protein 3, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP5882-ec-10ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15908429

  • 183.00€ / 10 microgramos


Fecha estimada de envío
20-08-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_003467.1
Cysteine and glycine-rich protein 1
10 μg
Not determined.
Full Length
Human
His-SUMO
Tris-based buffer, 50% glycerol
8048
Greater than 90% as determined by SDS-PAGE.
Research Use Only
CSRP3
Recombinant Protein
E. coli
MPNWGGGAKC GACEKTVYHA EEIQCNGRSF HKTCFHCMAC RKALDSTTVA AHESEIYCKV CYGRRYGPKG IGYGQGAGCL STDTGEHLGL QFQQSPKPAR SVTTSNPSKF TAKFGESEKC PRCGKSVYAA EKVMGGGKPW HKTCFRCAIC GKSLESTNVT DKDGELYCKV CYAKNFGPTG IGFGGLTQQV EKKE
Videos
Documentos