missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Cystathionine beta-synthase Protein

A DNA sequence encoding the Homo sapiens (Human) Cystathionine beta-synthase, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP5767-ec-1mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15917619

  • 1926.00€ / 1 miligramo


Fecha de envío estimada
del proveedor 05-04-2019
Añadir a la cesta

Especificación

Especificación

NP_000062.1
Cystathionine beta-synthase Protein
1 mg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
102724560
Greater than 90% as determined by SDS-PAGE.
Research Use Only
CBS
Recombinant Protein
E. coli
PSETPQAEVG PTGCPHRSGP HSAKGSLEKG SPEDKEAKEP LWIRPDAPSR CTWQLGRPAS ESPHHHTAPA KSPKILPDIL KKIGDTPMVR INKIGKKFGL KCELLAKCEF FNAGGSVKDR ISLRMIEDAE RDGTLKPGDT IIEPTSGNTG IGLALAAAVR GYRCIIVMPE KMSSEKVDVL RALGAEIVRT PTNARFDSPE SHVGVAWRLK NEIPNSHILD QYRNASNPLA HYDTTADEIL QQCDGKLDML VASVGTGGTI TGIARKLKEK CPGCRIIGVD PEGSILAEPE ELNQTEQTTY EVEGIGYDFI PTVLDRTVVD KWFKSNDEEA FTFARMLIAQ EGLLCGGSAG STVAVAVKAA QELQEGQRCV VILPDSVRNY MTKFLSDRWM LQKGFLKEED LTEKKPWWWH LRVQELGLSA PLTVLPTITC GHTIEILREK GFDQAPVVDE AGVILGMVTL GNMLSSLLAG KVQPSDQVGK VIYKQFKQIR LTDTLGRLSH ILEMDHFALV VHEQIQYHST GKSSQRQMVF GVVTAIDLLN FVAAQERDQK
Videos