missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Cyclin-K Protein

A DNA sequence encoding the Homo sapiens (Human) Cyclin-K, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP10034-ba-100ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15912788

  • 1210.00€ / 100 microgramos


Fecha estimada de envío
24-06-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_001092872.1
Cyclin-K Protein
100 μg
Not determined.
Extracellular Domain
Human
His
Tris-based buffer, 50% glycerol
8812
Greater than 90% as determined by SDS-PAGE.
Research Use Only
CCNK
Recombinant Protein
Baculovirus
MKENKENSSP SVTSANLDHT KPCWYWDKKD LAHTPSQLEG LDPATEARYR REGARFIFDV GTRLGLHYDT LATGIIYFHR FYMFHSFKQF PRYVTGACCL FLAGKVEETP KKCKDIIKTA RSLLNDVQFG QFGDDPKEEV MVLERILLQT IKFDLQVEHP YQFLLKYAKQ LKGDKNKIQK LVQMAWTFVN DSLCTTLSLQ WEPEIIAVAV MYLAGRLCKF EIQEWTSKPM YRRWWEQFVQ DVPVDVLEDI CHQILDLYSQ GKQQMPHHTP HQLQQPPSLQ PTPQVPQVQQ SQPSQSSEPS QPQQKDPQQP AQQQQPAQQP KKPSPQPSSP RQVKRAVVVS PKEENKAAEP PPPK
Videos