missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Cyclin D1 Protein

A DNA sequence encoding the Homo sapiens (Human) Cyclin D1, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP5776-ec-500ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15937679

  • 1180.00€ / 500 microgramos


Fecha estimada de envío
26-02-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_444284.1
Cyclin D1 Protein
500 μg
Not determined.
Full Length
Human
His
Tris-based buffer, 50% glycerol
595
Greater than 90% as determined by SDS-PAGE.
Research Use Only
CCND1
Recombinant Protein
E. coli
MEHQLLCCEV ETIRRAYPDA NLLNDRVLRA MLKAEETCAP SVSYFKCVQK EVLPSMRKIV ATWMLEVCEE QKCEEEVFPL AMNYLDRFLS LEPVKKSRLQ LLGATCMFVA SKMKETIPLT AEKLCIYTDN SIRPEELLQM ELLLVNKLKW NLAAMTPHDF IEHFLSKMPE AEENKQIIRK HAQTFVALCA TDVKFISNPP SMVAAGSVVA AVQGLNLRSP NNFLSYYRLT RFLSRVIKCD PDCLRACQEQ IEALLESSLR QAQQNMDPKA AEEEEEEEEE VDLACTPTDV RDVDI
Videos
Documentos