missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Cyclin D1 Protein
A DNA sequence encoding the Homo sapiens (Human) Cyclin D1, was expressed in the hosts and tags indicated.
Brand: enQuireBio™ QP5776-ec-200ug
Additional Details : Peso : 0.01000kg
Specifications
NP_444284.1 | |
Cyclin D1 Protein | |
Research Use Only | |
CCND1 | |
Recombinant Protein | |
E. coli | |
MEHQLLCCEV ETIRRAYPDA NLLNDRVLRA MLKAEETCAP SVSYFKCVQK EVLPSMRKIV ATWMLEVCEE QKCEEEVFPL AMNYLDRFLS LEPVKKSRLQ LLGATCMFVA SKMKETIPLT AEKLCIYTDN SIRPEELLQM ELLLVNKLKW NLAAMTPHDF IEHFLSKMPE AEENKQIIRK HAQTFVALCA TDVKFISNPP SMVAAGSVVA AVQGLNLRSP NNFLSYYRLT RFLSRVIKCD PDCLRACQEQ IEALLESSLR QAQQNMDPKA AEEEEEEEEE VDLACTPTDV RDVDI | |
Greater than 90% as determined by SDS-PAGE. |
595 | |
200 μg | |
Not determined. | |
Full Length | |
Human | |
His | |
Tris-based buffer, 50% glycerol |