missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CXCL3 / GRO gamma Protein

A DNA sequence encoding the Human CXCL3 / GRO gamma, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP10207-ec-250ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15933098

  • 1970.00€ / 250 microgramos


Fecha estimada de envío
19-03-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_002081.2
CXCL3 / GRO gamma Protein
250 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
ASVVTELRCQ CLQTLQGIHL KNIQSVNVRS PGPHCAQTEV IATLKNGKKA CLNPASPMVQ KIIEKILNKG STN
2921
Greater than 97% as determined by SDS-PAGE and HPLC.
Research Use Only
CXCL3
The biological activity determined by a chemotaxis bioassay using human CXCR2 transfected human 293 cells is in a concentration range of 10-100 ng/ml.
Human
Untagged
0.2 um filtered PBS, pH 7.4, lyophilized
Videos
Documentos