missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CXCL2 / MIP-2 Protein
Recombinant Protein
Marca: enQuireBio™ QP1016-100 ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_002080.1 | |
CXCL2 / MIP-2 Protein | |
Research Use Only | |
CXCL2 | |
Recombinant Protein | |
E. coli | |
APLATELRCQ CLQTLQGIHL KNIQSVKVKS PGPHCAQTEV IATLKNGQKA CLNPASPMVK KIIEKMLKNG KSN | |
Greater than 95% as determined by SDS-PAGE. |
2920 | |
100 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Human CXCL2 / MIP-2 induces Human neutrophil chemotaxis (significanty different from control at Human CXCL2 / MIP-2 concentrations greater than 10 ng/ml). | |
Human | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |