missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CXCL12 / SDF-1 Protein

Recombinant Protein

Marca:  enQuireBio™ QP5280-500ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15915839

  • 1972.00€ / 500 microgramos


Fecha estimada de envío
24-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_000600.1
CXCL12 / SDF-1 Protein
500 μg
Less than or equal to 1.00 EUs/ug as determined by the LAL method.
Human CXCL12 / SDF-1 induces primary human T cell chemotaxis (significanty different from control at Human CXCL12 / SDF-1 concentrations greater than 5-20 ng/ml).
Human
Untagged
Sterile filtered (0.22 um) solution containing 0.1% TFA
6387
Greater than 95% as determined by SDS-PAGE.
Research Use Only
CXCL12
Recombinant Protein
E. coli
KPVSLSYRCP CRFFESHVAR ANVKHLKILN TPNCALQIVA RLKNNNRQVC IDPKLKWIQE YLEKALNKRF KM
Videos