missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CXCL1 / MGSA / NAP-3 Protein
A DNA sequence encoding the Homo sapiens (Human) CXCL1 / MGSA / NAP-3, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP1010-ec-100ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001502.1 | |
CXCL1 / MGSA / NAP-3 Protein | |
Research Use Only | |
CXCL1 | |
Recombinant Protein | |
E. coli | |
ASVATELRCQ CLQTLQGIHP KNIQSVNVKS PGPHCAQTEV IATLKNGRKA CLNPASPIVK KIIEKMLNSD KSN | |
Greater than 90% as determined by SDS-PAGE. |
2919 | |
100 μg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |