Learn More
enQuireBio™ Recombinant Human CTGF Protein
Recombinant Protein
Marca: enQuireBio™ QP5373-20ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001892.1 | |
CTGF Protein | |
Research Use Only | |
CTGF | |
Recombinant Protein | |
E. coli | |
MGKKCIRTP K. ISKPIKFE LS. GCTSMKT YRA. KFCGVC TDGR. CCTPH RTTTL. PVEF KCPDGE. VMK KNMMFIK. TC ACHYNCPG. D NDIFESLYY. RKMYGDMA | |
Greater than 90 as determined by SDS-PAGE. |
1490 | |
20 μg | |
Less than or equal to 1.00 EUs/ug as determined by the LAL method. | |
Bioactivity of Human CTGF confirmed in HUVEC cell proliferation assay. | |
Human | |
Untagged | |
Sterile filtered (0.22 um) solution containing 0.1% TFA |