missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CTF1 Protein
A cDNA sequence encoding the CTF1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11552-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
1489 | |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Recombinant Protein | |
E. coli | |
MRGSHHHHHH GSSRREGSLE DPQTDSSVSL LPHLEAKIRQ THSLAHLLTK YAEQLLQEYV QLQGDPFGLPSFSPPRLPVA GLSAPAPSHA GLPVHERLRL DAAALAALPP LLDAVCRRQA ELNPRAPRLL RRLEDAARQA RALGAAVEAL LAALGAANRG PRAEPPAATA SAASATGVFP AKVLGLRVCG LYREWLSRTE GDLGQLLPGG SA | |
Purity of CTF1 Human Recombinant is greater than 90% as determined by SDS-PAGE. |
CTF1 Protein | |
Research Use Only | |
CTF1 | |
Human | |
His | |
CTF1 was filtered (0.4microm) and lyophilized from 0.5 mg/ml in 0.05M Acetate buffer pH 4. |