missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CST5 Protein
A cDNA sequence encoding the CST5 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP10562-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
CST5 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
QSRTLAGGIH ATDLNDKSVQ RALDFAISEY NKVINKDEYY SRPLQVMAAY QQIVGGVNYY FNVKFGRTTC TKSQPNLDNC PFNDQPKLKE EEFCSFQINE VPWEDKISIL NYKCRKVHHH HHH | |
Greater than 90.0% as determined by SDS-PAGE. |
1 mg | |
< 1.0 EU per ug protein as determined by the LAL method. | |
Human | |
His | |
The CST5 solution (0.5 mg/1ml) contains phosphate buffered saline (pH 7.4) and 10% glycerol. |