missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CSNK2A2 Protein

A cDNA sequence encoding the CSNK2A2 was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP11533-1mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15917558

  • 5941.00€ / 1 miligramo


Fecha estimada de envío
24-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

1459
Greater than 90% as determined by SDS-PAGE.
Research Use Only
CSNK2A2
Human
His
CSNK2A2 protein solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 8.0), 0.15M NaCl, 30% glycerol and 1mM DTT.
CSNK2A2 Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Recombinant Protein
E. coli
MGSSHHHHHH SSGLVPRGSH MGSHMPGPAA GSRARVYAEV NSLRSREYWD YEAHVPSWGN QDDYQLVRKL GRGKYSEVFE AINITNNERV VVKILKPVKK KKIKREVKIL ENLRGGTNII KLIDTVKDPV SKTPALVFEY INNTDFKQLY QILTDFDIRF YMYELLKALD YCHSKGIMHR DVKPHNVMID HQQKKLRLID WGLAEFYHPA QEYNVRVASR YFKGPELLVD YQMYDYSLDM WSLGCMLASM IFRREPFFHG QDNYDQLVRI AKVLGTEELY GYLKKYHIDL DPHFNDILGQ HSRKRWENFI HSENRHLVSP EALDLLDKLL RYDHQQRLTA KEAMEHPYFY PVVKEQSQPC ADNAVLSSGL TAAR
Videos