Learn More
enQuireBio™ Recombinant Human CRYBB1 Protein
A cDNA sequence encoding the CRYBB1 was constructed and used to recombinantly synthesize the protein.
Marca:  enQuireBio™ QP11520-5ug
Detalles adicionales : Peso : 0.01000kg
 Ver más versiones de este producto
Código de producto. 15917518
  • 137.00€ / 5 microgramos
Fecha estimada de envío: 28-05-2024
para ver el stock
Especificaciones
CRYBB1 Protein
Research Use Only
Recombinant Protein
E. coli
MSQAAKASAS ATVAVNPGPD TKGKGAPPAG TSPSPGTTLA PTTVPITSAK AAELPPGNYR LVVFELENFQ GRRAEFSGEC SNLADRGFDR VRSIIVSAGP WVAFEQSNFR GEMFILEKGE YPRWNTWSSS YRSDRLMSFR PIKMDAQEHK ISLFEGANFK GNTIEIQGDD APSLWVYGFS DRVGSVKVSS GTWVGYQYPG YRGYQYLLEP GDFRHWNEWG AFQPQMQSLR RLRDKQWHLE GSFPVLATEP PKRSHHHHHH
Greater than 90.0% as determined by SDS-PAGE.
5 μg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
His
The CRYBB1 solution (1 mg/ml) contains 20mM Tris-HCl pH 8.0, 1mM DTT and 10% glycerol.