missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human CRISP1 Protein
A cDNA sequence encoding the CRISP1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11505-10ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
CRISP1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MKHHHHHHAS KKKSARDQFN KLVTDLPNVQ EEIVNIHNAL RRRVVPPASN MLKMSWSEEA AQNARIFSKY CDMTESNPLE RRLPNTFCGE NMHMTSYPVS WSSVIGVWYS ESTSFKHGEW TTTDDDITTD HYTQIVWATS YLIGCAIASC RQQGSPRYLY VCHYCHEGND PETKNEPYKT GVPCEACPSN CEDKLCTNPC IYYDEYFDCD IQVHYLGCNH STTILFCKAT CLCDTEIK | |
Greater than 85% as determined by SDS-PAGE. |
10 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The protein was lyophilized from a 0.5 mg/ml solution containing 20mM TRIS, 50mM NaCl, pH 7.5. |