missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Cripto / TDGF1 Protein

A DNA sequence encoding the Homo sapiens (Human) Cripto / TDGF1, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP8703-ec-200ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16056041

  • 675.00€ / 200 microgramos


Fecha estimada de envío
26-02-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_001167607.1
Cripto / TDGF1 Protein
200 μg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
6997
Greater than 90% as determined by SDS-PAGE.
Research Use Only
TDGF1
Recombinant Protein
E. coli
GHQEFARPSR GYLAFRDDSI WPQEEPAIRP RSSQRVPPMG IQHSKELNRT CCLNGGTCML GSFCACPPSF YGRNCEHDVR KENCGSVPHD TWLPKKCSLC KCWHGQLRCF PQAFLPGCD
Videos
Documentos