missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Corticotropin-releasing factor receptor 1 Protein
A DNA sequence encoding the Homo sapiens (Human) Corticotropin-releasing factor receptor 1, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5867-ec-50ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001138618.1 | |
Corticotropin-releasing factor receptor 1 Protein | |
Research Use Only | |
CRHR1 | |
Recombinant Protein | |
E. coli | |
SLQDQHCESL SLASNISGLQ CNASVDLIGT CWPRSPAGQL VVRPCPAFFY GVRYNTTNNG YRECLANGSW AARVNYSECQ EILNEEKKSK VHYHVAVI | |
Greater than 90% as determined by SDS-PAGE. |
104909134 | |
50 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
GST | |
Tris-based buffer, 50% glycerol |