missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Corticotropin-releasing factor receptor 1 Protein

A DNA sequence encoding the Homo sapiens (Human) Corticotropin-releasing factor receptor 1, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP5867-ec-500ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15918329

  • 1210.00€ / 500 microgramos


Fecha estimada de envío
24-07-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_001138618.1
Corticotropin-releasing factor receptor 1 Protein
500 μg
Not determined.
Partial (see sequence information for more details).
Human
GST
Tris-based buffer, 50% glycerol
104909134
Greater than 90% as determined by SDS-PAGE.
Research Use Only
CRHR1
Recombinant Protein
E. coli
SLQDQHCESL SLASNISGLQ CNASVDLIGT CWPRSPAGQL VVRPCPAFFY GVRYNTTNNG YRECLANGSW AARVNYSECQ EILNEEKKSK VHYHVAVI
Videos