missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human COPZ1 Protein
A cDNA sequence encoding the COPZ1 was constructed and used to recombinantly synthesize the protein.
Marca: enQuireBio™ QP11487-2ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
COPZ1 Protein | |
Research Use Only | |
Recombinant Protein | |
E. coli | |
MGSSHHHHHH SSGLVPRGSH MEALILEPSL YTVKAILILD NDGDRLFAKY YDDTYPSVKE QKAFEKNIFN KTHRTDSEIA LLEGLTVVYK SSIDLYFYVI GSSYENELML MAVLNCLFDS LSQMLRKNVE KRALLENMEG LFLAVDEIVD GGVILESDPQ QVVHRVALRG EDVPLTEQTV SQVLQSAKEQ IKWSLLR | |
Greater than 95.0% as determined by SDS-PAGE. |
2 μg | |
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request. | |
Human | |
His | |
The COPZ1 solution (1 mg/1ml) contains 20mM Tris-HCl buffer (pH 8.0), 10% glycerol, 1mM DTT and 0.1M NaCl. |