missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Complement component C8 gamma chain Protein
A DNA sequence encoding the Homo sapiens (Human) Complement component C8 gamma chain, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP5739-ec-1mg
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_000597.2 | |
Complement component C8 gamma chain Protein | |
Research Use Only | |
C8G | |
Recombinant Protein | |
E. coli | |
QKPQRPRRPA SPISTIQPKA NFDAQQFAGT WLLVAVGSAC RFLQEQGHRA EATTLHVAPQ GTAMAVSTFR KLDGICWQVR QLYGDTGVLG RFLLQARDAR GAVHVVVAET DYQSFAVLYL ERAGQLSVKL YARSLPVSDS VLSGFEQRVQ EAHLTEDQIF YFPKYGFCEA ADQFHVLDEV RR | |
Greater than 90% as determined by SDS-PAGE. |
733 | |
1 mg | |
Not determined. | |
Full Length | |
Human | |
His-SUMO | |
Tris-based buffer, 50% glycerol |