missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Collagen alpha-1(XVIII) chain Protein
A DNA sequence encoding the Homo sapiens (Human) Collagen alpha-1(XVIII) chain, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP8749-ye-200ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_085059.2 | |
Collagen alpha-1(XVIII) chain Protein | |
Research Use Only | |
COL18A1 | |
Recombinant Protein | |
Yeast | |
QPVLHLVALN SPLSGGMRGI RGADFQCFQQ ARAVGLAGTF RAFLSSRLQD LYSIVRRADR AAVPIVNLKD ELLFPSWEAL FSGSEGPLKP GARIFSFDGK DVLRHPTWPQ KSVWHGSDPN GRRLTESYCE TWRTEAPSAT GQASSLLGGR LLGQSAASCH HAYIVLCIEN SFMTASK | |
Greater than 90% as determined by SDS-PAGE. |
80781 | |
200 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |