missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Collagen alpha-1(XV) chain Protein
A DNA sequence encoding the Human Collagen alpha-1(XV) chain, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP10332-ec-250ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_001846.3 | |
Collagen alpha-1(XV) chain Protein | |
Research Use Only | |
COL15A1 | |
The ED50 as determined by inhibiting the FGF basic-dependent proliferation of HUVEC migration is less than 2.0 ug/mL in the presence of Anti- Human Endostatin Polyclonal Antibody. | |
Human | |
Untagged | |
0.2 um filtered PBS, pH 7.4, lyophilized |
1306 | |
250 μg | |
Less than 1.0 EU/ug as determined by LAL method. | |
Partial (see sequence information for more details). | |
Recombinant Protein | |
E. coli | |
NYEKPALHLA ALNMPFSGDI RADFQCFKQA RAAGLLSTYR AFLSSHLQDL STIVRKAERY SLPIVNLKGQ VLFNNWDSIF SGHGGQFNMH IPIYSFDGRD IMTDPSWPQK VIWHGSSPHG VRLVDNYCEA WRTADTAVTG LASPLSTGKI LDQKAYSCAN RLIVLCIENS FMTDARK | |
Greater than 96% as determined by SDS-PAGE and HPLC. |