missing translation for 'onlineSavingsMsg'
Learn More
Learn More
enQuireBio™ Recombinant Human Coagulation Factor XI / FXI / F11 Protein
A DNA sequence encoding the Homo sapiens (Human) Coagulation Factor XI / FXI / F11, was expressed in the hosts and tags indicated.
Marca: enQuireBio™ QP6007-ec-500ug
Detalles adicionales : Peso : 0.01000kg
Especificaciones
NP_000119.1 | |
Coagulation Factor XI / FXI / F11 Protein | |
Research Use Only | |
F11 | |
Recombinant Protein | |
E. coli | |
ECVTQLLKDT CFEGGDITTV FTPSAKYCQV VCTYHPRCLL FTFTAESPSE DPTRWFTCVL KDSVTETLPR VNRTAAISGY SFKQCSHQIS ACNKDIYVDL DMKGINYNSS VAKSAQECQE RCTDDVHCHF FTYATRQFPS LEHRNICLLK HTQTGTPTRI TKLDKVVSGF SLKSCALSNL ACIRDIFPNT VFADSNIDSV MAPDAFVCGR ICTHHPGCLF FTFFSQEWPK ESQRNLCLLK TSESGLPSTR IKKSKALSGF SLQSCRHSIP VFCHSSFYHD TDFLGEELDI VAAKSHEACQ KLCTNAVRCQ FFTYTPAQAS CNEGKGKCYL KLSSNGSPTK ILHGRGGISG YTLRLCKMDN ECTTKIKPR | |
Greater than 90% as determined by SDS-PAGE. |
2160 | |
500 μg | |
Not determined. | |
Partial (see sequence information for more details). | |
Human | |
His | |
Tris-based buffer, 50% glycerol |