missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human Coagulation Factor VII / FVII / F7 Protein

A DNA sequence encoding the Homo sapiens (Human) Coagulation Factor VII / FVII / F7, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP6012-ec-200ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15929359

  • 985.00€ / 200 microgramos


Fecha estimada de envío
25-03-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_000122.1
Coagulation Factor VII / FVII / F7 Protein
200 μg
Not determined.
Partial (see sequence information for more details).
Human
His-SUMO
Tris-based buffer, 50% glycerol
2155
Greater than 90% as determined by SDS-PAGE.
Research Use Only
F7
Recombinant Protein
E. coli
ANAFLEELRP GSLERECKEE QCSFEEAREI FKDAERTKLF WISYSDGDQC ASSPCQNGGS CKDQLQSYIC FCLPAFEGRN CETHKDDQLI CVNENGGCEQ YCSDHTGTKR SCRCHEGYSL LADGVSCTPT VEYPCGKIPI LEKRNASKPQ GR
Videos
Documentos