missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CNTF Protein

A DNA sequence encoding the Human CNTF, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP10413-ec-100ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15933778

  • 668.00€ / 100 microgramos


Fecha estimada de envío
16-10-2019
Añadir a la cesta

Especificaciones

Especificaciones

NP_000605.1
CNTF Protein
100 μg
Less than 1.0 EU/ug as determined by LAL method.
Partial (see sequence information for more details).
Recombinant Protein
E. coli
MAFTEHSPLT PHRRDLCSRS IWLARKIRSD LTALTESYVK HQGLNKNINL DSADGMPVAS TDQWSELTEA ERLQENLQAY RTFHVLLARL LEDQQVHFTP TEGDFHQAIH TLLLQVAAFA YQIEELMILL EYKIPRNEAD GMPINVGDGG LFEKKLWGLK VLQELSQWTV RSIHDLRFIS SHQTGIPARG SHYIANNKKM
1270
Greater than 97% as determined by SDS-PAGE and HPLC.
Research Use Only
CNTF
The ED50 as determined by a cell proliferation assay using human TF-1 cells is less than 150 ng/ml, corresponding to a specific activity of > 6.7 M 103 IU/mg.
Human
Untagged
0.2 um filtered PBS, pH 7.4, lyophilized
Videos
Documentos