missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CNGA4 Protein

A DNA sequence encoding the Homo sapiens (Human) CNGA4, was expressed in the hosts and tags indicated.

Marca:  enQuireBio™ QP9966-iv-100ug

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 16049101

  • 2045.00€ / 100 microgramos


Fecha estimada de envío
26-02-2020
Añadir a la cesta

Especificaciones

Especificaciones

NP_001032406.1
CNGA4 Protein
100 μg
Not determined.
Full Length Transmembrane Protein
Human
His-SUMO
Tris-based buffer, 50% glycerol
1262
Greater than 85% as determined by SDS-PAGE.
Research Use Only
CNGA4
Recombinant Protein
Invitro-E. coli
MSQDTKVKTT ESSPPAPSKA RKLLPVLDPS GDYYYWWLNT MVFPVMYNLI ILVCRACFPD LQHGYLVAWL VLDYTSDLLY LLDMVVRFHT GFLEQGILVV DKGRISSRYV RTWSFFLDLA SLMPTDVVYV RLGPHTPTLR LNRFLRAPRL FEAFDRTETR TAYPNAFRIA KLMLYIFVVI HWNSCLYFAL SRYLGFGRDA WVYPDPAQPG FERLRRQYLY SFYFSTLILT TVGDTPPPAR EEEYLFMVGD FLLAVMGFAT IMGSMSSVIY NMNTADAAFY PDHALVKKYM KLQHVNRKLE RRVIDWYQHL QINKKMTNEV AILQHLPERL RAEVAVSVHL STLSRVQIFQ NCEASLLEEL VLKLQPQTYS PGEYVCRKGD IGQEMYIIRE GQLAVVADDG ITQYAVLGAG LYFGEISIIN IKGNMSGNRR TANIKSLGYS DLFCLSKEDL REVLSEYPQA QTIMEEKGRE ILLKMNKLDV NAEAAEIALQ EATESRLRGL DQQLDDLQTK FARLLAELES SALKIAYRIE RLEWQTREWP MPEDLAEADD EGEPEEGTSK DEEGRASQEG PPGPE
Videos
Documentos