missing translation for 'onlineSavingsMsg'
Learn More

enQuireBio™ Recombinant Human CLPP Protein

A cDNA sequence encoding the CLPP was constructed and used to recombinantly synthesize the protein.

Marca:  enQuireBio™ QP11451-1mg

Detalles adicionales : Peso : 0.01000kg

 Ver más versiones de este producto

Código de producto. 15937308

  • 6130.00€ / 1 miligramo


Fecha estimada de envío
20-03-2020
Añadir a la cesta

Especificaciones

Especificaciones

CLPP Protein
1 mg
Not Determined. Testing recommended prior to use in cell culture and can be performed upon request.
Human
Untagged
CLPP solution (0.5 mg/ml) containing 20mM Tris-HCl buffer (pH 7.5), 2mM DTT, 20% glycerol and 100mM NaCl.
Greater than 90.0% as determined by SDS-PAGE.
Research Use Only
Recombinant Protein
E. coli
MPLIPIVVEQ TGRGERAYDI YSRLLRERIV CVMGPIDDSV ASLVIAQLLF LQSESNKKPI HMYINSPGGV VTAGLAIYDT MQYILNPICT WCVGQAASMG SLLLAAGTPG MRHSLPNSRI MIHQPSGGAR GQATDIAIQA EEIMKLKKQL YNIYAKHTKQ SLQVIESAME RDRYMSPMEA QEFGILDKVL VHPPQDGEDE PTLVQKEPVE AAPAAEPVPA ST
Videos
Documentos